주요 문서
58442
Supelco
SSI LO-Pulse Damper
without cabinet
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
설명
316 stainless steel
cabinet no
특징
without cabinet
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
A pulse damper controls pump pulsations for a more stable baseline. The LO-Pulse damper is a patented unit compatible
with single piston reciprocating HPLC pumps (Altex 110A, Eldex pumps, LDC Mini-Pump VS, SSI Models 200 and 300,
etc.). At pressures from 500 psi to 6,000 psi (35-420 kg/cm2), it improves precision of quantitative analyses and detection limits for trace sample components. Fittings and instructions included.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
법적 정보
LO-Pulse is a trademark of Scientific Systems, Inc.
SSI is a trademark of Scientific Systems, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)