주요 문서
PHR9083
Supelco
5-(Phenylazo)salicylic acid
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
동의어(들):
Mesalazine impurity I (PhEur), 2-Hydroxy-5-(phenylazo)benzoic acid
로그인조직 및 계약 가격 보기
About This Item
실험식(Hill 표기법):
C13H10N2O3
CAS Number:
Molecular Weight:
242.23
UNSPSC 코드:
12352200
Grade
certified reference material
Quality Level
API family
mesalazine
제품 라인
TraceCERT®
grade
TraceCERT®
유통기한
limited shelf life, expiry date on the label
제조업체/상표
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
저장 온도
2-8°C
SMILES string
N(=Nc2ccccc2)c1cc(c(cc1)O)C(=O)O
InChI
1S/C13H10N2O3/c16-12-7-6-10(8-11(12)13(17)18)15-14-9-4-2-1-3-5-9/h1-8,16H,(H,17,18)
InChI key
JHDYSXXPQIFFJZ-UHFFFAOYSA-N
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
법적 정보
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
픽토그램
Exclamation mark
신호어
Warning
유해 및 위험 성명서
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)