About This Item
Recommended Products
isotopic purity
98 atom % D
Assay
97% (CP)
bp
105-110 °C (lit.)
mass shift
M+6
SMILES string
[2H]C([2H])([2H])CCNCCC([2H])([2H])[2H]
InChI
1S/C6H15N/c1-3-5-7-6-4-2/h7H,3-6H2,1-2H3/i1D3,2D3
InChI key
WEHWNAOGRSTTBQ-WFGJKAKNSA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
44.6 °F - closed cup
Flash Point(C)
7 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service