Skip to Content
Merck
All Photos(1)

Key Documents

938262

Sigma-Aldrich

Sodium modified silica particles

non-porous, 200 nm particle size

Synonym(s):

Sodium ion functionalized silica,, Sodium-functionalized silica nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352309

Quality Level

form

powder

particle size

200 nm

pore size

non-porous

bp

>2200 °C

mp

>1600 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Our sodium-modified silica particles are non-porous silica nanoparticles with a monodisperse particle size of 200 nm. The surface has been chemically modified to exhibit an increased concentration of sodium ions.

Application

Sodium functionalized silica particles can serve as an additive in electrolytes of sodium-ion batteries, particularly as fillers in hybrid liquid electrolytes or in polymer electrolytes. The functionalized particles can enhance the ionic conductivity of the electrolyte, improving the overall performance and stability of the battery. Sodium-modified silica particles can also find applications in drug delivery systems, diagnostics, or biosensing due to their enhanced surface functionality and potential for selective interactions with biological molecules.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service