Skip to Content
Merck
All Photos(2)

Key Documents

939641

Sigma-Aldrich

1-Methylindoline-2-thione

greener alternative

≥95%

Synonym(s):

1-Methyl-2,3-dihydro-1H-indole-2-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NS
CAS Number:
Molecular Weight:
163.24
MDL number:
UNSPSC Code:
12352100

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Photocatalysis

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to yellow

greener alternative category

SMILES string

S=C1N(C)C2=CC=CC=C2C1

InChI

InChI=1S/C9H9NS/c1-10-8-5-3-2-4-7(8)6-9(10)11/h2-5H,6H2,1H3

InChI key

GBSPZRWQIJCQQB-UHFFFAOYSA-N

General description

1-Methylindoline-2-thione is a indole thiolate often used in the synthesis of (Indoline-2-S) related products.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

1-Methylindoline-2-thione is a indole thiolate has been used in the synthesis of:
  • Brassilexin, Sinalexin and Wasalexins
  • Functionilized thiopyrano-indole annulated heterocycles
  • N-Alkylated Analogues of Indolylthio Glycosides
Also used as a photoreductant in:
  • The activation of C-F. C-Cl and C-O bonds
  • The synthesis of thioethers from aryl chlorides & alcohols

Features and Benefits

1-Methylindoline-2-thione is an affordable indole thiolate that is useful in general synthesis as well as the transition-metal free photocatalytic reduction of a range of strong polar C-F, C-O and C-Cl bonds.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service