941026
Riboflavin, 2′,3′,4′,5′-tetraacetate

>95%
Synonym(s):
(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)pentane-1,2,3,4-tetrayl tetraacetate, 2′,3′,4′,5′-tetraacetate, RFTA, Riboflavin, Riboflavin Tetraacetate
About This Item
solid
Recommended Products
Quality Level
Assay
>95%
form
powder or crystals
solid
reaction suitability
reagent type: catalyst
reaction type: Photocatalysis
storage condition
dry at room temperature
protect from light
color
light yellow to dark yellow
SMILES string
O=C(N1)C2=NC3=CC(C)=C(C)C=C3N(C[C@H](OC(C)=O)[C@@H]([C@H](OC(C)=O)COC(C)=O)OC(C)=O)C2=NC1=O
InChI
InChI=1S/C25H28N4O10/c1-11-7-17-18(8-12(11)2)29(23-21(26-17)24(34)28-25(35)27-23)9-19(37-14(4)31)22(39-16(6)33)20(38-15(5)32)10-36-13(3)30/h7-8,19-20,22H,9-10H2,1-6H3,(H,28,34,35)/t19-,20+,22-/m0/s1
InChI key
VKVDYPHLGLIXAG-VWPQPMDRSA-N
General description
Application
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service