Skip to Content
Merck

GF60703286

Polyethylene Terephthalate

(polyester, PET, PETP), film, thickness 0.075 mm, L 1 m, clear clear

Synonym(s):

ES301300

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352100
NACRES:
NA.23
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

film

manufacturer/tradename

Goodfellow 607-032-86

L

1 m

thickness

0.075 mm

color

clear clear

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service