Key Documents
TEM-CF300HCUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper Hexagonal Mesh Grid
size 300 mesh, box of 50
Synonym(s):
CARBON HEXAGONAL MESH - CU 300 MESH UL 50/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 50
size
300 mesh
General description
A 3-4 nm ultrathin film of pure carbon supported hexagonal mesh grids. Copper grid, 300 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)