Skip to Content
Merck
All Photos(1)

Key Documents

W408811

Sigma-Aldrich

trans-α-Damascone

new

≥90%

Synonym(s):

(±)-α-Damascone, (E)-1-(2,6,6-Trimethyl-1-cyclohex-2-enyl)but-2-en-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
FEMA Number:
4088
Beilstein:
2208707
MDL number:
UNSPSC Code:
12352200
Flavis number:
07.226
grade:
Halal
food allergen:
no known allergens

grade

Halal

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥90%

contains

IFRA restricted rose ketone

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; black currant; floral; fruity; rose

SMILES string

C\C=C\C(=O)C1C(C)=CCCC1(C)C

InChI key

CRIGTVCBMUKRSL-FNORWQNLSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F

Flash Point(C)

105 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service