Skip to Content
Merck
All Photos(1)

Key Documents

315M-TEW

Sigma-Aldrich

Anti-PD-1 (NAT105) Mouse Monoclonal Antibody

new

1mg/mL carrier free

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

NAT105, monoclonal

description

Purified mouse monoclonal antibody IgG1 in PBS without preservatives

species reactivity

human

packaging

vial of

concentration

1 mg/mL

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

Quality

Research Use Only (RUO)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service