Skip to Content
Merck
All Photos(1)

Key Documents

MABN638-I

Sigma-Aldrich

Anti-Amyloid beta fibrils Antibody, clone M87

clone M87

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

clone

M87, recombinant monoclonal

isotype

IgG

Specificity

Clone M87 is a conformation-sensitive rabbit recombinant monoclonal antibody that detects Amyloid beta fibrils. It recognizes a specific conformation of residues 3-7 of the amyloid beta peptide sequence (EFRHD) found in amyloid fibrils.

Immunogen

Fibrillar Abeta 1-42 peptide

Reconstitution

Please refer to lot specific datasheet.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service