Skip to Content
Merck
All Photos(1)

Key Documents

MABS2002

Sigma-Aldrich

Anti-mPEG, clone 15-2b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

mol wt

(N/A)

purified by

using Protein A

species reactivity

mouse,

isotype

IgG2b, kappa

Specificity

Clone 15-2b specifically detects proteins conjugated with methoxy polyethylene glycol (mPEG).

Application

Anti-mPEG, clone 15-2b, Cat. No. MABS2002, is a mouse monoclonal antibody that detects methoxy-PEG (mPEG)-conjugated proteins and has been tested for use in ELISA.
ELISA

Features and Benefits

Methoxy PEG covalently conjugated to E.coli beta-glucuronidase.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Purified mouse monoclonal antibody IgG2b in PBS with 0.04% sodium azide.

Other Notes

Polyethylene glycols (PEG) are non-toxic, uncharged, hydrophilic long chain polymers with a big exclusion volume in aqueous solutions. The covalent attachment of PEG is commonly employed to modify proteins and drugs that can increase their bioavailability and reduce immunogenicity and proteolytic degradation. The monomethoxylated form of polyethylene glycol (mPEG) has been widely employed in drug modifications. mPEG modification reduces nonspecific binding and prevents recognition by the reticuloendothelial system. This monoclonal antibody (clone 15-2b) specifically binds to the terminal methoxy group, but not the backbone of PEG. It displays high affinity for the terminal methoxy group and can be easily used as a detection antibody for the quantification of mPEG by ELISA. It is also suitable for detection of PEGylated liposomes and nanoparticles. The mPEG cell-based competition ELISA can detect lower molecular-weight (less than 2 kDa) mPEG molecules even at nanomolar concentration and the assay is not affected by the presence of mouse or human serum. In addition, mPEG cell-based ELISA can be useful in determining the pharmacokinetics of a mPEG molecule an animal without the need for any radiolabeling
Evaluated by ELISA with methoxy Polyethylene Glycol (10KPEG, 3KPEG, and 2K mPEG).

ELISA Analysis: 5 µg/mL of this antibody detected methoxy polyethylene glycol (2K-mPEG) and with minimal reactivity with 10K and 3K amino-PEGs.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service