Skip to Content
Merck
All Photos(1)

Key Documents

MABT1560

Sigma-Aldrich

Anti-CD9 Antibody, clone 4.1F12

clone 4.1F12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

clone

4.1F12, monoclonal

isotype

IgG2cκ

Specificity

Clone 4.1F12 is a rat monoclonal antibody that detects murine CD9.

Immunogen

Mouse platelets fused with X63 mouse myeloma.

Reconstitution

Please refer to lot specific datasheet.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service