MABT883
Anti-CD315 Antibody, clone 17H1
clone 17H1, from rat
Synonym(s):
CD9 partner 1, CD9P1, Glu-Trp-Ile EWI motif-containing protein F, nEWI-F, Prostaglandin F2-alpha receptor regulatory protein, Prostaglandin F2-alpha receptor-associated protein
About This Item
Recommended Products
biological source
rat
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
17H1, monoclonal
species reactivity
human
technique(s)
flow cytometry: suitable
immunohistochemistry: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... PTGFRN(5738)
General description
Specificity
Immunogen
Application
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected CD315 in Squamous cell carcinoma tissue (Courtesy of Reinhard Zeidler, Helmholtz Zentrum Muenchen, Neuherberg, Germany in cooperation with Dr. A. Bankfalvi, Inst. für Pathologie Universitätsklinkum, Essen).
Cell Structure
Quality
Flow Cytometry Analysis: 1 µg of this antibody detected CD315 in one million A549 cells.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service