Skip to Content
Merck
All Photos(1)

Key Documents

P2B010A05G1

Millipore

Pellicon 2 Biomax 10 kD 0.5m2 with Winged Gasket

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806

Quality Level

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service