NIST1721
Southern oceanic air
NIST® SRM® 1721
Synonym(s):
Compression air
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of cylinder
application(s)
pharmaceutical (small molecule)
format
mixture
InChI
1S/C37H49N3O5S.2ClH/c1-27(2)37(36(41)38(3)30-14-8-10-16-34(30)46-37)29-13-7-9-15-31(29)45-24-12-11-18-39-20-22-40(23-21-39)19-17-28-25-32(42-4)35(44-6)33(26-28)43-5;;/h7-10,13-16,25-27H,11-12,17-24H2,1-6H3;2*1H/t37-;;/m1../s1
InChI key
HQTMLRUQTHILKM-CHKASDEDSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service