Skip to Content
Merck
All Photos(1)

Key Documents

NIST3113

Cobalt (Co) standard solution

NIST® SRM® 3113

Synonym(s):

Cobalt (Co) Standard Solution, Cobalt nitrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

single component solution

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of cobalt. A unit of SRM 3113 consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of cobalt. The solution contains nitric acid at a volume fraction of approximately 10 %. For more information, please refer to the SDS and COA.

SRM 3113_cert SRM 3113 _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service