Skip to Content
Merck
All Photos(1)

Key Documents

NIST3135A

Neodymium (Nd) Standard Solution

new

NIST®SRM® 3135A

Synonym(s):

Neodymium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

packaging

pkg of 5 × 10 mL

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of neodymium. SRM 3135A_cert SRM 3135A _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service