Skip to Content
Merck
All Photos(1)

Key Documents

SMB01393

Sigma-Aldrich

Medetomidine hydrochloride

new

≥95%

Synonym(s):

5-[1-(2,3-dimethylphenyl)ethyl]-1H-imidazole;hydrochloride, Domitor, Medetomidine HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17ClN2
CAS Number:
Molecular Weight:
236.74
UNSPSC Code:
12352200
form:
powder or crystals
solubility:
DMF: soluble
DMSO: soluble
PBS: soluble
ethanol: soluble
water: soluble

biological source

synthetic

Quality Level

description

Antibody Research Area: Analgesic, Sedative

Assay

≥95%

form

powder or crystals

color

white to off-white

mp

168-172 °C

solubility

DMF: soluble
DMSO: soluble
PBS: soluble
ethanol: soluble
water: soluble

storage temp.

−20°C

SMILES string

CC(C1=C(C(C)=CC=C1)C)C2=CN=CN2.Cl

InChI

InChI=1S/C13H16N2.ClH/c1-9-5-4-6-12(10(9)2)11(3)13-7-14-8-15-13;/h4-8,11H,1-3H3,(H,14,15);1H

InChI key

VPNGEIHDPSLNMU-UHFFFAOYSA-N

General description

Medetomidine Hydrochloride, is a It is a synthetic product and known to have α2 adrenergic agonist activity. It is often used as anesthetic, sedative and analgesic.

Application

High purity product for studying anesthetic effect.

Features and Benefits

Versatile and adaptable for wide variety of research applications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service