Skip to Content
Merck
All Photos(1)

Key Documents

BAH378480002

Scienceware® velveteen squares for replica plater

size 6 in. × 6 in., sterile, black cotton, pkg of 36 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

black cotton

sterility

sterile

packaging

pkg of 36 ea

manufacturer/tradename

Bel-Art H37848-0002

size

6 in. × 6 in.

Looking for similar products? Visit Product Comparison Guide

General description

Dimensions: 152mm (6" x 6"). Packed in a sealed bag with 3 inner bags that can be resealed. Each bag holds 12 squares.
Sterile, 100% cotton material
  • Highly porous, twill-lined velveteen squares
  • Use to easily and rapidly reproduce cell colonies
  • Choose white or black colors for better contrast with cells being replicated

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service