Skip to Content
Merck
All Photos(1)

Key Documents

BMSMS1000

Benchmark SmartSeal Semi-Automated Microplate Sealer

new

AC/DC input 115 V, US 2-pin plug

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

feature

48 mm maximum plate height
Sealing time - 0.5 seconds to 10 seconds (0.1 second increments)

manufacturer/tradename

Benchmark Scientific MS1000

parameter

temp. range at 80-200 °C

width × depth × H

-17.8 cm -7 in. -37 cm -14.6 in. -33 cm -13 in.

Looking for similar products? Visit Product Comparison Guide

General description

The Accuris SmartSeal Automated Plate Sealer provides fast, reproducible sealing of microplates. Unlike manual plate sealing, the SmartSeal ensures uniform sealing of all wells and consistency from one plate to the next. With digital control over all parameters (time, temperature and pressure) the SmartSeal is capable of accepting a wide range of plate types and heights. An aluminum block adapter is included (for assay and PCR plates) increasing the compatibility of the instrument to allow for sealing nearly any plate up to 48mm tall. With sealing times as low as 5 seconds, the SmartSeal is a time saving solution for labs and production facilities requiring high quality and consistent plate sealing.

Compatible with:
Deep well (storage) plates up to 48mm tall
Assay plates
PCR Plates (skirterd, semi-skirted and non-skirted)
Polypropylene polystyrene or Polyethylene plates
Foil or polymer (transparent) seals

Application

Heat sealing microplates for storage and qPCR.

Features and Benefits

Works with plates up to 48mm tall Adjustable pressure, temperature and time to achieve ideal sealing conditions for a variety of different films and plates

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service