Key Documents
BMSNS1000EUK
Benchmark SmartDrop L Nano Spectrophotometer
new
AC/DC input 230 V (GB plug)
About This Item
UNSPSC Code:
12352200
manufacturer/tradename
Benchmark Scientific NS1000-E-UK
load volume
1.0-2.0 μL
General description
The Accuris SmartDrop series of nano spectrophotometers provides quick and easy quantification of small samples. Utilizing a small sample volume eliminates the need for dilutions, providing more accurate results. A full color, 7” touchscreen with icon-driven menus and preprogrammed protocols gets users up and running quickly. No external computer is required. Each unit features a quartz pedestal onto which a drop of sample is placed. The arm is lowered and the drop is stretched into a liquid column of specificed length for spectral analysis. DNA, RNA and protein concentrations and purity numbers are displayed in a matter of seconds. The resulting data can be printed using the integrated printer, saved in the SmartDrop’s onboard memory or exported via USB as an Excel file.
Application
Spectrophotometry, OD600
Features and Benefits
  • Quick and accurate results from a sample droplet
  • Touch screen control panel, no PC or software required
  • Built-in printer and cuvette port for OD600 readings
Choose from one of the most recent versions:
Certificates of Analysis (COA)