Skip to Content
Merck
All Photos(1)

Key Documents

BHK21-Hektor

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

hamster kidney

packaging

tube of 5 μg 05062301-DNA-5UG
pkg of vial of cells 05062301-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Syrian kidney, serum-free

Cell Line Description

BHK21CL13-2P cell line (Sigma Catalogue number. 84111301) adapted to grow in the chemically defined protein- and peptide-free Hektor G medium.

Culture Medium

Hektor G medium + 4mM Glutamine + 5 ug/ml Insulin. When freezing cells down use Hektor G medium + 4mM Glutamine (50:50 conditioned medium:fresh medium) + 10% DMSO.

Subculture Routine

Viability may be poor on resuscitation and may initially decrease further. Full recovery may take up to 2 weeks. A centrifugation step to remove the cryoprotectant is essential. Rapidly thaw the frozen ampoule in a water bath at 37°C for 1-2 minutes. Tran

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

Hektor is a trademark of Cell Culture Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service