Skip to Content
Merck
All Photos(1)

Key Documents

F9061

SAFC

ISF-1 Medium

with 2.438 g/L sodium bicarbonate, with L-alanyl-L-glutamine, with pluronic F-68, without antibiotics

Synonym(s):

Cell culture medium, Culture medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

form

liquid

Quality Level

quality

with 2.438 g/L sodium bicarbonate, with L-alanyl-L-glutamine, with pluronic F-68, without antibiotics

packaging

pkg of 1 L

shipped in

ambient

storage temp.

2-8°C

Application

Serum-free, low-protein medium formulated to support the growth of hybridomas used in the manufacture of monoclonal antibodies.

Features and Benefits

This product is identical to Biochrom GmbH part number F9061-01. ISF-1 is designed to support the rapid growth of cells to high densities and the maintenance of viable cells for extended culture periods. This formulation reduces raw material cost and the low protein level reduces cost associated with downstream processing. Purified components in the product reduce the variability sometimes encountered with biological materials leading to improved consistency of the medium performance. Cells can be successfully transferred from serum-supplemented cultures to this medium with little or no adaptation.

Caution

Do not freeze. Protect from light. Several components of the medium are light labile and should not be exposed to light for lengthy periods of time. When stored at 2-8 °C in the dark, medium that has not been supplemented is stable until the expiration date on the label.

Other Notes

This proprietary formulation includes inorganic salts, essential and non-essential amino acids, stable glutamine, vitamins, sodium bicarbonate, trace elements, fatty acids, and other organics. It contains low concentrations of recombinant human insulin, bovine serum albumin and human transferrin (source tested negative for HIV antibody and HbsAG).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service