Skip to Content
Merck
All Photos(3)

Key Documents

PB10100702CN

Pop-Bio Vü-C Chemilumisnescence Imaging System

Synonym(s):

Imaging system, Pop-Bio Vü-C Chemilumisnescence Imaging System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600

AC/DC input

24 V

packaging

pkg of 1 ea

Looking for similar products? Visit Product Comparison Guide

General description

Using Advanced Progressive Imaging technology, the high sensitivity sensors can easily capture low output blots to give you an outstanding image. All you have to do is to select the tray size for your blot, place the blot in the marked position and push the tray into Vü with your finger. Vü immediately recognizes the tray and then adapts to the blot size or application type. You will get your image in a matter of a few minutes.

Vü is a stunning new system which utilizes a new imaging platform. It sits discreetly on any laboratory bench. Just use your cell phone, tablet, laptop or computer to collect the image and that′s all you have to do. Depending on your sample type, your image will appear in seconds or minutes ready for you to print or analyze. No fuss, no hassle, no drama, no camera, lens or filters to worry about. No sensitivity issues, no focusing needed.

Application

Western blots

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service