Skip to Content
Merck
All Photos(3)

Key Documents

PB10100704EU

Pop-Bio Vü-F Fluorescence Imaging System

Synonym(s):

Imaging system, Pop-Bio Vü-F Fluorescence Imaging System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600

AC/DC input

24 V, EuroPlug

packaging

pkg of 1 ea

Looking for similar products? Visit Product Comparison Guide

General description

Vü is for fluorescence applications such as DNA gels, protein gels, safe dye gels. The sliding drawer can accept samples up to 20cm x 20cm and has a series of trays available for different application types. Just place your sample on the tray and push it in. Vü detects the sample type and captures an image using either UV, blue light. The use of traditional ethidium bromide stained UV gels may be declining but Vü is still built with a 302nm light source. For safe dyes there are integral blue LED′s. You can still use a traditional white light converter to view commassie and silver stain samples.

With Vü you can take any mobile device, tablet, laptop, computer and link it with your Vü. In fact, your only control is to capture the image – no fussy setups are required because we don′t use a camera, lens or filter wheel. Just push the sample draw in and Vü does the rest.

Application

Fluorescent gels

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service