Skip to Content
Merck
All Photos(1)

Key Documents

SAB5900009

Alpaca Anti-Mouse single domain antibody, agarose beads

new

recombinant Alpaca(VHH) Nano antibody agarose bead suspension

Sign Into View Organizational & Contract Pricing


About This Item

biological source

alpaca

Quality Level

recombinant

expressed in Pichia pastoris

conjugate

agarose conjugate

antibody product type

primary antibodies

clone

recombinant monoclonal

form

liquid

species reactivity

mouse

technique(s)

ChIP: suitable
RIP: suitable
co-immunoprecipitation (co-IP): suitable
immunoprecipitation (IP): suitable
mass spectrometry (MS): suitable

color

colorless

matrix

(~45 um (cross-linked to 7.5% agarose beads))

isotype

VHH

capacity

binding capacity(1.0 mL Anti-Mouse single domain antibody, agarose beads slurry binds to ~4 mg mouse IgG)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-Mouse Agarose VHH Beads are suspension of activated agarose beads coupled with anti-mouse IgG nanobodies. This product is suitable for pulling down protein that is coupled with mouse IgGs or has mouse Fc tag.

Specificity

Binds to mouse IgG/mouse Fc tag

Immunogen

Mouse IgG

Features and Benefits

  • Anti Tag-Nano antibody beads are single-domain/variable region of heavy chain of camelid antibodies(sdAb/VHH).
  • They are directly coupled to beads covalently.
  • They have unique properties such as small size (~15kDa, 2-4nm) when compared to conventional antibodies.
  • No heavy and light chains in downstream interactions.
  • Acids and alkali resistance, tolerance to harsh conditions.
  • Short incubation time 5 – 30 minutes.
  • Low batch to batch variations.
  • Typically, antibodies are not detectable in Western Blot.

Packaging

plastic tube

Physical form

1XPBS, 0.03% sodium azide, 50% glycerol.

Other Notes

Before usage, please invert the vial several times (DO NOT VORTEX) to form beads suspension, and take out beads’ suspension using a pipette tip with bigger opening (i.e., cut the tip of a 1000 μl pipette tip with sterile scissors). Once opened, it is recommended to use paraffin film to seal the cap.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service