Skip to Content
Merck
All Photos(2)

Key Documents

SML3873

Sigma-Aldrich

Pip6 PARP-1 inhibitor

≥98% (HPLC)

Synonym(s):

(S)-4-(3-(3-(1,1-difluoroethyl)-6-methyl-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine-7-carbonyl)-4-fluorobenzyl)phthalazin-1(2H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21F3N6O2
CAS Number:
Molecular Weight:
482.46
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cell penetrant and potent inhibitor of PARP-1 that exhibits potent cytotoxicity in cancer cells.


Pip6 is a cell penetrant and potent inhibitor of PARP-1 (Poly (ADP-ribose) polymerase 1) that exhibits potent cytotoxicity in cancer cells. It appears that Pip6 improved cytotoxicity is due to enhanced target residence time on PARP-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service