Skip to Content
Merck
All Photos(2)

Key Documents

SML3968

Sigma-Aldrich

NSI-189

≥98% (HPLC)

Synonym(s):

(4-Benzylpiperazin-1-yl)-[2-(3-methylbutylamino)pyridin-3-yl]methanone, NSI 189, [2-[(3-Methylbutyl)amino]-3-pyridinyl][4-(phenylmethyl)-1-piperazinyl]methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N4O
CAS Number:
Molecular Weight:
366.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N2(CCN(CC2)C(=O)c3c(nccc3)NCCC(C)C)Cc1ccccc1

InChI

1S/C22H30N4O/c1-18(2)10-12-24-21-20(9-6-11-23-21)22(27)26-15-13-25(14-16-26)17-19-7-4-3-5-8-19/h3-9,11,18H,10,12-17H2,1-2H3,(H,23,24)

InChI key

DYTOQURYRYYNOR-UHFFFAOYSA-N

Biochem/physiol Actions

Orally available neurogenic drug that ameliorates impaired memory function in Zucker diabetic fatty rats; exhibits long-acting antidepressant, procognitive, synaptoplastic, and neurotrophic activities.


NSI-189 is an orally available neurogenic drug with pleiotropic properties that ameliorates impaired memory function and plasticity in Zucker diabetic fatty rat model of type 2 diabetes through enhanced expression of mitochondrial complexes I, III, IV, and V. NSI-189 exhibits long-acting antidepressant, procognitive, synaptoplastic, and neurotrophic activities in patients with major depressive disorder.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service