Skip to Content
Merck
All Photos(1)

Key Documents

SML4082

Paliperidone Palmitate

new

≥98% (HPLC)

Synonym(s):

3-(2-(4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl)ethyl)-2-methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-9-yl palmitate, 9-Hydroxyrisperidone palmitate, Hexadecanoic acid 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl ester, RO 92670

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H57FN4O4
CAS Number:
Molecular Weight:
664.89
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

soluble (DMSO:TFA, 2 mg/mL, clear (add 1 drop TFA))

storage temp.

-10 to -25°C

SMILES string

Fc1cc2[o]nc(c2cc1)C3CCN(CC3)CCC4=C(NC5=C(CCCN5C4=O)OC(=O)CCCCCCCCCCCCCCC)C

InChI key

YOSKWMGLLZWACB-UHFFFAOYSA-N

Biochem/physiol Actions

Prodrug form of paliperidone, is a long-acting injectable antipsychotic that targets dopamine and serotonin receptors.


Paliperidone palmitate is a long-acting injectable form of paliperidone, an atypical antipsychotic that targets dopamine D2 and serotonin 5-HT2A receptors to alleviate schizophrenia symptoms through neurotransmission regulation. Paliperidone palmitate also interacts with α1/α2 adrenergic and H1 histaminergic receptors, influencing both its therapeutic effects and side effect profile. The formulation′s sustained release mechanism promotes gradual absorption, ensuring stable drug levels for prolonged efficacy.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service