Skip to Content
Merck

ZRB2127

Sigma-Aldrich

Anti-KIF5B Antibody, clone 2F23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Kinesin-1 heavy chain;Conventional kinesin heavy chain;Ubiquitous kinesin heavy chain;UKHC

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F23, recombinant monoclonal

description

2F23 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 109.69 kDa
observed mol wt ~115 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

porcine, feline, bovine, canine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... KIF5B(3799)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2F23 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Kinesin-1 heavy chain (KIF5B). It targets an epitope within 16 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Kinesin-1 heavy chain (KIF5B).

Application

Quality Control Testing

Evaluated by Western Blotting in C2C12 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected KIF5B in C2C12 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected KIF5B in Human brain and Mouse brain tissue lysates.

Affinity Binding Assay: A representative lot of this antibody bound KIF5B peptide with a KD of 4.6 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected KIF5B in HT-29 and U2OS cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected KIF5B in human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in C2C12 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected KIF5B in C2C12 cell lysate.

Target description

Kinesin-1 heavy chain (UniProt: P33176; also known as Conventional kinesin heavy chain, Ubiquitous kinesin heavy chain, UKHC, KIF5B) is encoded by the KIF5B (also known as KNS, KNS1) gene (Gene ID: 3799) in human. Kinesin-1 is an abundant, ATP-dependent motor protein that moves along microtubule filaments. It participates in a number of important intracellular transport processes. Kinesin is a heterotetramer, comprised of 2 heavy chains and 2 light chains. The heavy chains dimerize to form an elongated stalk with 2 amino-terminal globular domains (motor domains) at one end and the light chains at the other end. Kinesin can pull a cellular cargo along a microtubule, slide one microtubule relative to another, or even remodel the microtubule cytoskeleton through regulation of microtubule dynamics. It plays a key role in mitosis and in the trafficking of organelles and vesicles. The Kinesin heavy chain is shown to move its cytoplasmic cargo toward the plus ends of microtubules. KIF5B is ubiquitous in its distribution and is shown to be uniformly distributed between soma and neurites in hippocampal neurons. It participates in the normal distribution of mitochondria and lysosomes in the cell and can induce formation of neurite-like membrane protrusions in non-neuronal cells in a Zinc Finger FYVE-Type Containing 27 (ZFYVE27)-dependent manner. It also regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle, it is reported to antagonize dynein function and drives the separation of nuclei and centrosomes. It is also required for anterograde axonal transportation of mitogen-activated protein kinase 8 interacting protein 3 (MAPK8IP3) and is essential for MAPK8IP3 function in axon elongation. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service