D2622KRSUA
Sulfur in Odourless Kerosene, ASTM D2622
Calibration Set A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
manufacturer/tradename
Paragon Scientific Ltd
application(s)
petroleum
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
Components
Set A
Blank, 0.0005 % (5ppm), 0.0010 % (10ppm), 0.0100 % (100ppm), 0.0250 % (250ppm), 0.0500 % (500ppm), 0.0750 % (750ppm), 0.1000 % (1000ppm). 100 mL ea.
Blank, 0.0005 % (5ppm), 0.0010 % (10ppm), 0.0100 % (100ppm), 0.0250 % (250ppm), 0.0500 % (500ppm), 0.0750 % (750ppm), 0.1000 % (1000ppm). 100 mL ea.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service