PHL10012
3-Feruloyl-4-caffeoylquinic acid
phyproof® Reference Substance
Synonym(s):
3-Feruloyl-4-caffeoylquinic acid
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
technique(s)
HPLC: suitable
application(s)
food and beverages
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
OC1=CC(/C=C/C(O[C@@H]2[C@H](O)C[C@@](O)(C(O)=O)C[C@H]2OC(/C=C/C3=CC(OC)=C(O)C=C3)=O)=O)=CC=C1O
InChI key
SDMADMBVKYOYQN-CSPRBKSFSA-N
Related Categories
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service