Skip to Content
Merck
Dihydropinene BioRenewable, reagent grade

Dihydropinene

Dihydropinene BioRenewable, reagent grade
Synonym(s):
Pinane, 2,6,6-Trimethylbicyclo[3.1.1]heptane
Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25

Products

Brand
Sustainability
Color
Feature
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing