
Lithium nickel manganese cobalt oxide

Empirical Formula (Hill Notation):
LiNi0.5Mn0.3Co0.2O2
CAS Number:
Molecular Weight:
96.55
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing