Skip to Content
Merck
All Photos(1)

Key Documents

939579

Sigma-Aldrich

Lithium nickel manganese cobalt oxide

new

lithium-niobate coated NMC532 cathode active material

Synonym(s):

LiNbO3-coated NMC532, NMC532

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
LiNi0.5Mn0.3Co0.2O2
CAS Number:
Molecular Weight:
96.55
UNSPSC Code:
26111700

grade

battery grade

Quality Level

description

application(s): battery manufacturing

Assay

99% trace metals basis

form

powder

mol wt

96.56 g/mol

composition

LiNi0.5Mn0.3Co0.2O2

density

4.77 g/cm3 ((lit.))

Looking for similar products? Visit Product Comparison Guide

General description

Lithium niobate coated NMC532 is a powdered form of lithium nickel manganese cobalt oxide with a stoichiometry of 5:3:2 Ni:Mn:Co and a typical particle size of 4-10 microns. The particles are coated in 5-10 nm amorphous lithium niobate, less than 1 wt.%.

Application

LiNbO3-coated NMC532 is a high-performance cathode active material designed to be used in lithium-ion batteries (LIBs), particularly solid-state batteries and high-voltage LIBs. NMC532 balances the high capacity bestowed by 50% nickel with the structural stability imparted by the manganese and cobalt. Compared to NMC811, NMC532 tends to achieve better cycling lifetime and coulombic efficiencies. The LiNbO3-coating boosts the stability of the cathode material by protecting the electrode and electrolyte from unwanted reactions. Research indicates that the lithium-niobate coating, increases the capacity over time (less capacity fade) by mitigating reactions between the electrolyte and cathode material and by impeding the release of metal ions from the NMC cathode into the electrolyte. Additionally, the lithium-niobate coating, sometimes called a LiNbO3 insertion layer, improves the rate capability by reducing the space charge layer barrier at the electrolyte/cathode interface. This improvement is evidenced both in enhanced Li diffusion and smaller electrode polarization.

Features and Benefits

-NMC532 balances high capacity and long cycle lifetimes
-LiNbO3 coating protects the cathode and electrolyte, improving coulombic efficiency
-LiNbO3 insertion layer improves rate capability

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service