Merck
  • Biocompatible chiral monolithic stationary phase synthesized via atom transfer radical polymerization for high performance liquid chromatographic analysis.
Biocompatible chiral monolithic stationary phase synthesized via atom transfer radical polymerization for high performance liquid chromatographic analysis.
Journal of chromatography. A (2015-07-23)
Huai-Song Wang, Xia-Yi Feng, Ji-Ping Wei
ABSTRACT
Novel biocompatible chiral monolithic stationary phase was prepared by reverse and direct atom transfer radical polymerization (ATRP) methods. By taking advantages of the controlled/living property of ATRP method, the chiral monolith was prepared by reverse ATRP (RATRP) firstly. An attractive feature of RATRP is the prepared polymer containing a terminal radically transferable atom that can initiate another post-polymerization reaction by direct ATRP. Then, the biocompatible poly(hydroxyethyl methacrylate) (PHEMA) was grafted on the surface of the chiral monolith by direct ATRP as a diffusion barrier for proteins. This biocompatible chiral monolith was successfully used as restricted access stationary phase for determination of enantiomers in biological samples with direct injection by high-performance liquid chromatography (HPLC).
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
2-Hydroxyethyl methacrylate
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Diethylenetriamine, ReagentPlus®, 99%
Sigma-Aldrich
Methacryloyl chloride, technical grade, 90%
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%
Sigma-Aldrich
Methacryloyl chloride, 97%, contains monomethyl ether hydroquinone as stabilizer