Skip to Content
Merck

COMH97F0EDE0

Combi-Blocks, Inc.

7'-Fluorospiro[cyclopropane-1,3'-indoline]

Sign Into View Organizational & Contract Pricing

About This Item

CAS Number:
Molecular Weight:
163.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

7'-fluorospiro[cyclopropane-1,3'-indoline]

Inchi Code

1S/C10H10FN/c11-8-3-1-2-7-9(8)12-6-10(7)4-5-10/h1-3,12H,4-6H2

InChI key

UHRUFLKWQXZJJM-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QK-9027

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QK-9027


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library