Skip to Content
MilliporeSigma

AABH99A4CA11

AA BLOCKS, INC.

2-((1H-Benzo[d]imidazol-1-yl)methyl)-6-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

Sign Into View Organizational & Contract Pricing

About This Item

CAS Number:
Molecular Weight:
290.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

2-((1H-benzo[d]imidazol-1-yl)methyl)-6-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

Inchi Code

1S/C17H14N4O/c1-12-5-4-8-16-19-13(9-17(22)21(12)16)10-20-11-18-14-6-2-3-7-15(14)20/h2-9,11H,10H2,1H3

InChI key

ICTBPKWPCGSFPZ-UHFFFAOYSA-N

storage temp.

2~8C

Purity

98%

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/1434288-24-0

Synonym

2-(1H-BENZIMIDAZOL-1-YLMETHYL)-6-METHYL-4H-PYRIDO[1,2-A]PYRIMIDIN-4-ONE

MSDS

https://www.aablocks.com/downloadSds?cas=1434288-24-0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library