AATB-5502
AAT Bioquest
AAT Bioquest ReadiLink™ KLH Conjugation Kit
2 Reactions
About This Item
Recommended Products
manufacturer/tradename
AAT Bioquest
General description
This ReadiLink™ KLH Conjugation kit is primarily optimized for the simple preparation of hapten-carrier conjugates for immunization and antibody production. The ReadiLink™ KLH Conjugation kit is one-step conjugation of a hapten to a carrier protein using the carboxyl-reactive carbodiimide as the crosslinker. The resulting conjugate is used for eliciting an immune response and antibody production against the hapten.
Components:
- EDC (1-ethy 1-3-[dimethylaminopropyl] carbodiimide hydrochloride)-2 X 10 mg
- Conjugation Buffer (pH 4.7)-1 bottle (20 mL)
- mcKLH-2 X 2 mg
- Purification Buffer Salts (pH 7.2)-2 X 10 mL
- Spin Desalting Columns (7K MWCO)-2 X 2 mL
ReadiLink™ KLH Conjugation Kit is manufactured by AAT Bioquest
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.