Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900916

Sigma-Aldrich

N-Azidoacetylglucosamine-tetraacylated

95%

Synonym(s):

Ac4GlcNAz

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N4O10
CAS Number:
Molecular Weight:
430.37
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

assay

95%

form

powder or crystals

availability

available only in USA

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

N-Azidoacetylglucosamine-tetraacylated (Ac4GlcNAz) provides the first part of a simple and robust two-step technique that helps identify and characterize cell surface sialic acid-containing glycoproteins. The azide-modified protein can be detected by reaction with alkynes. For example alkynes labeled with a fluorescent probe or a biotin can be used. The acetyl groups increase cell permeability and allow the unnatural sugars to easily pass through the cell membrane. Carboxyesterases remove the acetyl groups once the monosaccharide is in the cell.

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Self-react. C

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service