Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

938432

Sigma-Aldrich

EZ-Link Biotin-BMCC

≥95%

Synonym(s):

(3aS,4S,6aR)-N-[4-[[[4-[(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)methyl]cyclohexyl]carbonyl]amino]butyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide, 1-Biotinamido-4-(4′-[maleimidomethyl-cyclohexane]-carboxamido)butane, BMCC, Biotin-BMCC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H39N5O5S
CAS Number:
Molecular Weight:
533.68
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21

Quality Level

assay

≥95%

form

powder or crystals

color

white to light brown

storage temp.

2-8°C

SMILES string

O=C(NCCCCNC(C1CCC(CN2C(C=CC2=O)=O)CC1)=O)CCCC[C@@H]3SC[C@]([C@]3([H])N4)([H])NC4=O

InChI

InChI=1S/C26H39N5O5S/c32-21(6-2-1-5-20-24-19(16-37-20)29-26(36)30-24)27-13-3-4-14-28-25(35)18-9-7-17(8-10-18)15-31-22(33)11-12-23(31)34/h11-12,17-20,24H,1-10,13-16H2,(H,27,32)(H,28,35)(H2,29,30,36)/t17?,18?,19-,20-,24-/m0/s1

InChI key

ZKTPOUXCUHYLDZ-SAYNGHPTSA-N

Application

Biotin BMCC is heterobifunctional crosslinking reagent useful for conjugation to lysine residues in proteins. Biotin handle can be used for affinity chromatography of labelled proteins/peptides

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service