Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

939366

Sigma-Aldrich

Nickel Citrate hydrate

new

99.9% trace metals basis

Synonym(s):

Nickel(II) citrate hydrate, Trinickel dicitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni3C12H10O14 · xH2O
CAS Number:
Molecular Weight:
554.28 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
assay:
>98% (by complexometric EDTA)
99.9% trace metals basis
form:
powder or crystals
solubility:
water: soluble

type

(High purity Salts)

Quality Level

assay

>98% (by complexometric EDTA)
99.9% trace metals basis

form

powder or crystals

color

faint green to very dark green

solubility

water: soluble

anion traces

sulfate (SO42-): ≤50 ppm

cation traces

Al: <100 ppm
Cu: <100 ppm
Fe: <100 ppm
K: <100 ppm
Mg: <100 ppm
Na: ≤50 ppm
Pb: <100 ppm
Zn: <100 ppm

SMILES string

[Ni+2].[Ni+2].[Ni+2].[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-].[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-].O

InChI key

AXGFXGXJPPCFJX-UHFFFAOYSA-H

General description

Nickel acetate hydrate is an organic acid salt. Nickel citrate hydrate is a green crystalline solid that is soluble in water. This compound is commonly used in various applications such as electroplating, catalysts, and as a precursor in the synthesis of other nickel-containing compounds. In electroplating processes, nickel citrate hydrate acts as a source of nickel ions for depositing a layer of nickel onto surfaces.

Application

Nickel citrate hydrate can be used as a Nickel precursor

  • To synthesize these catalysts, the nickel precursor solution was impregnated onto the surface of the AlMCM-41 support. The impregnated support was then subjected to suitable drying and calcination processes to remove any solvent or excess precursor and activate the catalyst. The catalyst prepared with nickel citrate demonstrated stronger metal-support interaction, higher metal dispersion, and superior catalytic activity compared to those prepared with nickel nitrate and alkaline tetraamine nickel nitrate. The enhanced metal-acid interaction and (de)hydrogenating capability of this catalyst resulted in the highest catalytic activity and isomerization selectivity in n-dodecane hydroconversion.

  • In the synthesis of LaNiO3 perovskite, the Pechini method is employed due to its superior capabilities in achieving precise stoichiometric control, excellent reproducibility, and homogeneous product formation. This technique outperforms physical methods in ensuring the desired composition and quality of the LaNiO3 perovskite material.

Features and Benefits

  • Water soluble
  • Medium purity (99.9%)
  • Low trace metals in ppm level
  • Cost effective
  • Low Chloride and sulfate levels

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Met. Corr. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

target_organs

Respiratory Tract

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service