941522
Dextran methacrylate

30% degree of methacrylation
Synonym(s):
Dex-MA, DexMa, GeIMA, Methacrylate, Methacrylate-functionalized dextran, Methacrylated dextran, Methacrylic Anhydride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
powder or chunks (or Fibres)
color
white to faint yellow, faint brown
storage temp.
2-8°C
Related Categories
General description
Dextran methacrylate is a versatile natural polymer with a wide range of biomedical applications. Derived from non-toxic and hydrophilic polysaccharide, dextran, this polymer offers remarkable biocompatibility, biodegradability, and biological activity. The added methacrylate functionalization expands its potential, enabling photopolymerization to form crosslinked scaffolds and hydrogels for 3D cell encapsulation, cell culture, tissue engineering, and regenerative medicine.
Application
Dextran methacrylate can be used in a wide range of biomedical research applications including:
- Scaffold fabrication - dextran methacrylate can be photopolymerized to create crosslinked scaffolds, providing a 3D environment for cell encapsulation and tissue regeneration
- Tissue engineering - the hydrogel formed from dextran methacrylate promotes cell growth and differentiation within a 3D matrix
- Drug delivery - dextran methacrylate can be used to encapsulate and delivery drugs and therapeutic cells in a controlled and sustained manner
Features and Benefits
- Biocompatible and biodegradable
- Photopolymerizable
- 30-50% functionalized
- Tailorable for tethering additional functional groups or biochemical moieties or adjusting 3D hydrogel biochemical or mechanical properites
related product
Product No.
Description
Pricing
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service