Key Documents
BAH369650000
SP Bel-Art® Sterileware Plastic Sampling Knife
white polypropylene, L 20.9 cm, sterile, pack of 200 ea (individually wrapped)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
white polypropylene
sterility
sterile
packaging
pack of 200 ea (individually wrapped)
manufacturer/tradename
Bel-Art H36965-0000
L
20.9 cm
Overall L
8.25 in. (209 mm)
General description
Ideal for cutting small samples from fibrous pieces, this sterile sampler knife has a blade that is serrated on one edge and smooth on the other.
Features and Benefits
  • Serrated edge is capable of cutting seafood, meat, poultry, vegetables and other fibrous materials
  • FDA grade polypropylene plastic, safe for food, drug and cosmetic use
  • Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)