Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR734674

BRAND® robotic filter tips

new

PP, 40-1,000 µl, sterile, TipRack, BIO-CERT® LIQUID HANDLING QUALITY, 960EA

Synonym(s):

40-1000 ul, 734674, BRAND tips, TipRack, filter tips, microtiter plate, pipette tips, robotic tips, sterile tips, tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
volume:
40—1000 μL
sterility:
sterile
feature:
ATP free
PCR clean
RNase and DNase free
endotoxin free
filter
pyrogen free
packaging:
pack of 960 ea
manufacturer/tradename:
BRAND 734674

material

polypropylene tip

sterility

sterile

feature

ATP free
PCR clean
RNase and DNase free
endotoxin free
filter
pyrogen free

packaging

pack of 960 ea

manufacturer/tradename

BRAND 734674

volume

40—1000 μL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

For use with pipetting robots, tips are required that are matched to the robot′s mode of operation in terms of geometry and surface properties. BRAND® robotic pipette tips and robotic filter tips meet all requirements for use with the BRAND® Liquid Handling Station pipetting robot. The raw materials are free of DiHEMDA and Oleamid. Tips are manufactured at BRAND® under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used.

Features and Benefits

  • BIO-CERT® LIQUID HANDLING QUALITY: Free of DNA, RNase, DNase, Pyrogens, PCR-Inhibitors and ATP
  • For use with the BRAND® Liquid Handling Station pipetting robot

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service