GF71213284
Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8
500mm tube, 1.96mm outside diameter, 0.37mm wall thickness, 1.22mm inside diameter
Synonym(s):
NI027200
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
form
tubes
manufacturer/tradename
Goodfellow 712-132-84
L × wall thickness
500 mm × 0.37 mm
O.D. × I.D.
1.96 mm × 1.22 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service