Key Documents
Z679216
Reusable laboratory coat
size XXL
About This Item
UNSPSC Code:
46181532
size
XXL
feature
CE compliant
availability
not available in USA and Canada
General description
Heavyweight 245 g/m, polyester/cotton material with hydrofoil finish, giving Type 6 limited chemical splash protection.
  • Concealed stud fastening
  • Two hip pockets, one top pocket and side trouser pocket access
  • Finished back length 112 cm with centre back vent
Conforms to prEN1513:1997
Choose from one of the most recent versions:
Certificates of Analysis (COA)