Key Documents
FT1010
Alkali Scientific™
Alkali Scientific™ PurePoint™ Low Retention Filter Tips, Universal Fit
100 μL, Extra-Long, 960/Pk (10 racks of 96)
Synonym(s):
filter pipette tips, pipetting tips, pipettor tips, filter tips, Pipette tips, pipette tips, pipet tips
About This Item
PDF Icon
Shipped & Sold By Alkali Scientific
A Trusted Marketplace Seller
PDF Icon
Have a question?
binding type
low binding surface
feature
graduations: Yes
volume
0.01 mL
10 μL
manufacturer/tradename
Alkali Scientific™
General description
PurePoint™ Aerosol Barrier Filter Pipet Tips are designed and manufactured using premium-grade 100% polypropylene for the most critical molecular biology liquid handling applications. All PurePoint™ Aerosol Filter Tips are gamma irradiated and Certified RNase/DNase-Free, human DNA-Free, and Pyrogen-Free. In addition, PurePoint™ Aerosol Filter Tips are Ultra-Clear with a Fine-Point design for the convenient, precise pipetting of precious nucleic acid and protein samples and are designed to be Universal-fit for the majority of research-grade pipettors.
  • Universal-fit for most single and multichannel pipettors*
  • Gamma-irradiated and Graduated
  • Certified RNase, DNase, Human DNA, Pyrogen-FREE
  • Low-Retention, Fine-Point design for lowest sample adhesion
  • Ultra-Clear polymer withstands organic solvents and provides clear, error-free pipetting
  • Effective hydrophobic aerosol barrier eliminates carry-over and pipettor contamination
  • 10/Racks of 96/ 960/Pack
*Not suitable for Rainin LTS (Light Touch) Pipettors.
Legal Information
PurePoint™ is a trademark of Alkali Scientific.
Choose from one of the most recent versions:
Certificates of Analysis (COA)