TPN1006-NT
Alkali Scientific™
Alkali Scientific™ CellPro™ Non-Treated Cell Culture Plates
6 Well , Flat,Non-Treated, 1/pk, 50/cs
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Alkali Scientific™
General description
* High clarity, 100% virgin polystyrene
* Vacuum Plasma tissue culture treatment
* Clear lot number for batch tractability
* Peel-to-open, individually wrapped & sterilized by gamma irradiation
* Markings of well coordinates available for 12, 24, 48 and 96 well plates
* Non-Pyrogenic, DNase/RNase FREE
Legal Information
CellPro™ is a trademark of Alkali Scientific.
Choose from one of the most recent versions:
Certificates of Analysis (COA)